Assay Details
| Gene Symbol: Lamtor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502922.2 | XP_003502970.1 | 1 - 2 | 174 | 78 | |
| XM_007643537.1 | XP_007641727.1 | 1 - 2 | 174 | 78 | ||
| GenBank mRNA | JI867134.1 | - | 1 - 2 | 116 | 78 | |
| JP056109.1 | - | 1 - 2 | 116 | 78 | ||
More Information
Set Membership: |
|

Lamtor2
Genomic Map