Assay Details
| Gene Symbol: Ccdc88b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515020.1 | XP_003515068.1 | 7 - 8 | 666 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc88b| Gene Symbol: Ccdc88b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515020.1 | XP_003515068.1 | 7 - 8 | 666 | 58 | |
Set Membership: |
|