Assay Details
Gene Symbol: Cstb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515067.2 | XP_003515115.1 | 2 - 3 | 296 | 71 | |
GenBank mRNA | JI866087.1 | - | 2 - 3 | 290 | 71 | |
JP060891.1 | - | 2 - 3 | 290 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
