Assay Details
Gene Symbol: Ube2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640763.1 | XP_007638953.1 | 3 - 4 | 208 | 76 | |
GenBank mRNA | JI886082.1 | - | 1 - 2 | 91 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
