Assay Details
| Gene Symbol: Socs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640350.1 | XP_007638540.1 | 9 - 10 | 1393 | 60 | |
| GenBank mRNA | JI872925.1 | - | 1 - 2 | 145 | 60 | |
| JP048750.1 | - | 1 - 2 | 145 | 60 | ||
More Information
Set Membership: |
|

Socs7
Genomic Map