Assay Details
| Gene Symbol: Dnaaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654210.1 | XP_007652400.1 | 2 - 3 | 872 | 93 | |
| GenBank mRNA | JI881438.1 | - | 1 - 2 | 641 | 93 | |
More Information
Set Membership: |
|

Dnaaf2
Genomic Map