Assay Details
| Gene Symbol: LOC100768872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495305.2 | XP_003495353.1 | 5 - 6 | 550 | 104 | |
| GenBank mRNA | JI873675.1 | - | 5 - 6 | 547 | 104 | |
More Information
Set Membership: |
|

LOC100768872
Genomic Map