Assay Details
| Gene Symbol: Ube2z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503938.2 | XP_003503986.1 | 5 - 6 | 611 | 66 | |
| GenBank mRNA | JI878653.1 | - | 5 - 6 | 611 | 66 | |
| JP044270.1 | - | 5 - 6 | 611 | 66 | ||
More Information
Set Membership: |
|

Ube2z
Genomic Map