Assay Details
Gene Symbol: Suox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506194.2 | XP_003506242.1 | 3 - 4 | 310 | 75 | |
GenBank mRNA | JI895337.1 | - | 2 - 3 | 248 | 75 | |
JP048233.1 | - | 2 - 3 | 248 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
