Assay Details
Gene Symbol: Banf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509438.2 | XP_003509486.1 | 2 - 3 | 189 | 100 | |
XM_007650033.1 | XP_007648223.1 | 2 - 3 | 201 | 100 | ||
XM_007650034.1 | XP_007648224.1 | 2 - 3 | 153 | 100 | ||
XM_007650035.1 | XP_007648225.1 | 2 - 3 | 152 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
