Assay Details
Gene Symbol: LOC100772476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509461.2 | XP_003509509.1 | 2 - 3 | 406 | 95 | |
GenBank mRNA | JI878921.1 | - | 2 - 3 | 324 | 95 | |
JP039710.1 | - | 2 - 3 | 335 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
