Assay Details
Gene Symbol: Itm2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514035.2 | XP_003514083.2 | 2 - 3 | 559 | 68 | |
GenBank mRNA | JI869424.1 | - | 2 - 3 | 559 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
