Assay Details
Gene Symbol: LOC100758725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498260.2 | XP_003498308.2 | 1 - 2 | 800 | 77 | |
GenBank mRNA | JI877307.1 | - | 1 - 2 | 241 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
