Assay Details
Gene Symbol: LOC100767953 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503218.2 | XP_003503266.2 | 10 - 11 | 1138 | 90 | |
XM_003503219.2 | XP_003503267.1 | 10 - 11 | 1738 | 90 | ||
XM_007643826.1 | XP_007642016.1 | 9 - 10 | 1108 | 90 | ||
XM_007643827.1 | XP_007642017.1 | 11 - 12 | 1152 | 90 | ||
XM_007643828.1 | XP_007642018.1 | 11 - 12 | 1131 | 90 | ||
XM_007643829.1 | XP_007642019.1 | 9 - 10 | 3597 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
