Assay Details
Gene Symbol: LOC100767953 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503218.2 | XP_003503266.2 | 5 - 6 | 508 | 93 | |
XM_003503219.2 | XP_003503267.1 | 5 - 6 | 1108 | 93 | ||
XM_007643826.1 | XP_007642016.1 | 5 - 6 | 508 | 93 | ||
XM_007643827.1 | XP_007642017.1 | 6 - 7 | 522 | 93 | ||
XM_007643828.1 | XP_007642018.1 | 6 - 7 | 501 | 93 | ||
XM_007643829.1 | XP_007642019.1 | 4 - 5 | 2967 | 93 | ||
GenBank mRNA | JI878443.1 | - | 3 - 4 | 307 | 93 | |
JP045029.1 | - | 3 - 4 | 307 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
