Assay Details
| Gene Symbol: LOC100767953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503218.2 | XP_003503266.2 | 6 - 7 | 644 | 66 | |
| XM_003503219.2 | XP_003503267.1 | 6 - 7 | 1244 | 66 | ||
| XM_007643826.1 | XP_007642016.1 | 6 - 7 | 644 | 66 | ||
| XM_007643827.1 | XP_007642017.1 | 7 - 8 | 658 | 66 | ||
| XM_007643828.1 | XP_007642018.1 | 7 - 8 | 637 | 66 | ||
| XM_007643829.1 | XP_007642019.1 | 5 - 6 | 3103 | 66 | ||
| GenBank mRNA | JI878443.1 | - | 4 - 5 | 443 | 66 | |
| JP045029.1 | - | 4 - 5 | 443 | 66 | ||
More Information
Set Membership: |
|

LOC100767953
Genomic Map