Assay Details
| Gene Symbol: LOC100771702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506272.2 | XP_003506320.1 | 14 - 15 | 1205 | 138 | |
| XM_007646760.1 | XP_007644950.1 | 13 - 14 | 1087 | 138 | ||
| XM_007646761.1 | XP_007644951.1 | 14 - 15 | 1159 | 138 | ||
| XM_007646762.1 | XP_007644952.1 | 14 - 15 | 1237 | 138 | ||
| GenBank mRNA | JI870764.1 | - | 14 - 15 | 1203 | 138 | |
| JP037358.1 | - | 14 - 15 | 1203 | 138 | ||
More Information
Set Membership: |
|

LOC100771702
Genomic Map