Assay Details
| Gene Symbol: Rph3al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506715.2 | XP_003506763.1 | 7 - 8 | 862 | 60 | |
| XM_007647255.1 | XP_007645445.1 | 8 - 9 | 875 | 60 | ||
| XM_007647256.1 | XP_007645446.1 | 8 - 9 | 846 | 60 | ||
| XM_007647257.1 | XP_007645447.1 | 7 - 8 | 828 | 60 | ||
| XM_007647258.1 | XP_007645448.1 | 7 - 8 | 806 | 60 | ||
| XM_007647259.1 | XP_007645449.1 | 7 - 8 | 788 | 60 | ||
| GenBank mRNA | JI884792.1 | - | 2 - 3 | 196 | 60 | |
| JP054544.1 | - | 2 - 3 | 196 | 60 | ||
More Information
Set Membership: |
|

Rph3al
Genomic Map