Assay Details
Gene Symbol: LOC100771607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507606.2 | XP_003507654.1 | 2 - 3 | 735 | 59 | |
XM_007648204.1 | XP_007646394.1 | 2 - 3 | 738 | 59 | ||
GenBank mRNA | JI893254.1 | - | 1 - 2 | 350 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
