Assay Details
Gene Symbol: LOC100771607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507606.2 | XP_003507654.1 | 6 - 7 | 1299 | 72 | |
XM_007648204.1 | XP_007646394.1 | 6 - 7 | 1302 | 72 | ||
GenBank mRNA | JI878790.1 | - | 2 - 3 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
