Assay Details
Gene Symbol: Oraov1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510067.2 | XP_003510115.1 | 1 - 2 | 172 | 88 | |
XM_007650710.1 | XP_007648900.1 | 1 - 2 | 178 | 88 | ||
XM_007650711.1 | XP_007648901.1 | 1 - 2 | 179 | 88 | ||
XM_007650712.1 | XP_007648902.1 | 1 - 2 | 178 | 88 | ||
GenBank mRNA | JI868183.1 | - | 1 - 2 | 144 | 88 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
