Assay Details
Gene Symbol: Oraov1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510067.2 | XP_003510115.1 | 2 - 3 | 281 | 71 | |
XM_007650710.1 | XP_007648900.1 | 2 - 3 | 287 | 71 | ||
XM_007650711.1 | XP_007648901.1 | 2 - 3 | 288 | 71 | ||
XM_007650712.1 | XP_007648902.1 | 2 - 3 | 287 | 71 | ||
XM_007650713.1 | XP_007648903.1 | 2 - 3 | 353 | 71 | ||
GenBank mRNA | JI868183.1 | - | 2 - 3 | 253 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
