Assay Details
| Gene Symbol: Etaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642998.1 | XP_007641188.1 | 4 - 5 | 2322 | 95 | |
| GenBank mRNA | JI896490.1 | - | 1 - 2 | 1911 | 95 | |
More Information
Set Membership: |
|

Etaa1
Genomic Map