Assay Details
| Gene Symbol: Dtx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648099.1 | XP_007646289.1 | 4 - 5 | 1595 | 90 | |
| XM_007648100.1 | XP_007646290.1 | 4 - 5 | 1614 | 90 | ||
| XM_007648101.1 | XP_007646291.1 | 3 - 4 | 881 | 90 | ||
| XM_007648102.1 | XP_007646292.1 | 4 - 5 | 1586 | 90 | ||
More Information
Set Membership: |
|

Dtx3
Genomic Map