Assay Details
Gene Symbol: Chrna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497926.2 | XP_003497974.1 | 1 - 2 | 61 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
