Assay Details
Gene Symbol: Rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498617.2 | XP_003498665.1 | 3 - 4 | 270 | 70 | |
XM_007630611.1 | XP_007628801.1 | 3 - 4 | 267 | 70 | ||
GenBank mRNA | JP063188.1 | - | 3 - 4 | 240 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
