Assay Details
Gene Symbol: Nr3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500702.2 | XP_003500750.1 | 8 - 9 | 2938 | 74 | |
XM_007641448.1 | XP_007639638.1 | 9 - 10 | 3032 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
