Assay Details
Gene Symbol: Ndufb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507028.2 | XP_003507076.1 | 2 - 3 | 378 | 82 | |
GenBank mRNA | JI894126.1 | - | 2 - 3 | 402 | 82 | |
JP057672.1 | - | 2 - 3 | 402 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
