Assay Details
Gene Symbol: Tspan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512512.2 | XP_003512560.1 | 5 - 6 | 639 | 64 | |
GenBank mRNA | JI873960.1 | - | 4 - 5 | 438 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
