Assay Details
Gene Symbol: LOC100752430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643734.1 | XP_007641924.1 | 2 - 3 | 695 | 64 | |
GenBank mRNA | JI882993.1 | - | 2 - 3 | 425 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
