Assay Details
Gene Symbol: Ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007627424.1 | XP_007625614.1 | 2 - 3 | 337 | 75 | |
GenBank mRNA | JI865872.1 | - | 2 - 3 | 349 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
