Assay Details
| Gene Symbol: LOC100757102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499154.2 | XP_003499202.1 | 4 - 5 | 475 | 64 | |
| GenBank mRNA | JI883024.1 | - | 1 - 2 | 76 | 64 | |
| JP048185.1 | - | 1 - 2 | 76 | 64 | ||
More Information
Set Membership: |
|

LOC100757102
Genomic Map