Assay Details
| Gene Symbol: Snrpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508115.2 | XP_003508163.1 | 4 - 5 | 426 | 65 | |
| XM_007648739.1 | XP_007646929.1 | 4 - 5 | 538 | 65 | ||
| GenBank mRNA | JP060896.1 | - | 4 - 5 | 461 | 65 | |
More Information
Set Membership: |
|

Snrpb2
Genomic Map