Assay Details
Gene Symbol: Csrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496467.2 | XP_003496515.2 | 3 - 4 | 519 | 89 | |
XM_007650434.1 | XP_007648624.1 | 3 - 4 | 246 | 89 | ||
GenBank mRNA | JI894900.1 | - | 3 - 4 | 294 | 89 |
