Assay Details
Gene Symbol: Csrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496467.2 | XP_003496515.2 | 5 - 6 | 1124 | 86 | |
XM_007650434.1 | XP_007648624.1 | 5 - 6 | 851 | 86 | ||
GenBank mRNA | JI889220.1 | - | 1 - 2 | 277 | 86 |
