Assay Details
Gene Symbol: Clpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501766.2 | XP_003501814.1 | 4 - 5 | 592 | 61 | |
GenBank mRNA | JI896459.1 | - | 4 - 5 | 620 | 61 | |
JP061864.1 | - | 4 - 5 | 620 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
