Assay Details
| Gene Symbol: Paqr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509053.2 | XP_003509101.1 | 3 - 4 | 511 | 70 | |
| XM_007649673.1 | XP_007647863.1 | 3 - 4 | 511 | 70 | ||
| GenBank mRNA | JI891566.1 | - | 3 - 4 | 454 | 70 | |
| JP057900.1 | - | 3 - 4 | 454 | 70 | ||
More Information
Set Membership: |
|

Paqr3
Genomic Map