Assay Details
Gene Symbol: Aurkaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509971.2 | XP_003510019.1 | 3 - 4 | 549 | 59 | |
XM_007650637.1 | XP_007648827.1 | 3 - 4 | 660 | 59 | ||
GenBank mRNA | JI877400.1 | - | 1 - 2 | 374 | 59 | |
JP060205.1 | - | 1 - 2 | 377 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
