Assay Details
| Gene Symbol: Unc119b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644142.1 | XP_007642332.1 | 5 - 6 | 726 | 60 | |
| GenBank mRNA | JI888146.1 | - | 4 - 5 | 519 | 60 | |
More Information
Set Membership: |
|

Unc119b
Genomic Map