Assay Details
| Gene Symbol: Ube2u | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007614502.1 | XP_007612692.1 | 5 - 6 | 470 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2u| Gene Symbol: Ube2u | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007614502.1 | XP_007612692.1 | 5 - 6 | 470 | 62 | |
Set Membership: |
|