Assay Details
| Gene Symbol: LOC100751498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652831.1 | XP_007651021.1 | 7 - 8 | 908 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100751498| Gene Symbol: LOC100751498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652831.1 | XP_007651021.1 | 7 - 8 | 908 | 100 | |
Set Membership: |
|