Assay Details
| Gene Symbol: Otulin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641806.1 | XP_007639996.1 | 8 - 9 | 838 | 67 | |
| GenBank mRNA | JI873142.1 | - | 4 - 5 | 405 | 67 | |
More Information
Set Membership: |
|

Otulin
Genomic Map