Assay Details
| Gene Symbol: Asb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496120.2 | XP_003496168.2 | 3 - 4 | 307 | 73 | |
| XM_007647016.1 | XP_007645206.1 | 3 - 4 | 331 | 73 | ||
| XM_007647021.1 | XP_007645211.1 | 3 - 4 | 310 | 73 | ||
More Information
Set Membership: |
|

Asb14
Genomic Map