Assay Details
Gene Symbol: Cdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496417.2 | XP_003496465.1 | 1 - 2 | 301 | 64 | |
XM_007649925.1 | XP_007648115.1 | 1 - 2 | 301 | 64 | ||
GenBank mRNA | JI863265.1 | - | 1 - 2 | 222 | 64 | |
JP050650.1 | - | 1 - 2 | 222 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
