Assay Details
| Gene Symbol: Apoa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506866.2 | XP_003506914.1 | 2 - 3 | 144 | 57 | |
| XM_007647403.1 | XP_007645593.1 | 3 - 4 | 847 | 57 | ||
| XM_007647404.1 | XP_007645594.1 | 3 - 4 | 978 | 57 | ||
More Information
Set Membership: |
|

Apoa2
Genomic Map