Assay Details
Gene Symbol: Magohb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510551.2 | XP_003510599.1 | 3 - 4 | 330 | 66 | |
GenBank mRNA | JP038976.1 | - | 3 - 4 | 327 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
