Assay Details
| Gene Symbol: Rap1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512255.2 | XP_003512303.1 | 4 - 5 | 343 | 78 | |
| GenBank mRNA | JI873802.1 | - | 4 - 5 | 391 | 78 | |
| JP065854.1 | - | 4 - 5 | 391 | 78 | ||
More Information
Set Membership: |
|

Rap1a
Genomic Map