Assay Details
| Gene Symbol: Pcyox1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649241.1 | XP_007647431.1 | 4 - 5 | 880 | 62 | |
| GenBank mRNA | JI883697.1 | - | 1 - 2 | 221 | 62 | |
| JP055540.1 | - | 1 - 2 | 221 | 62 | ||
More Information
Set Membership: |
|

Pcyox1l
Genomic Map