Assay Details
| Gene Symbol: Txndc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496498.2 | XP_003496546.1 | 1 - 2 | 556 | 74 | |
| GenBank mRNA | JP064387.1 | - | 1 - 2 | 216 | 74 | |
More Information
Set Membership: |
|

Txndc12
Genomic Map