Assay Details
Gene Symbol: Elac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506798.2 | XP_003506846.1 | 3 - 4 | 646 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
